  • Search Results
  • Sodium butyrate protects against high-fat diet-induced oxidative stress in rat liver by promoting expression of nuclear factor E2-related factor 2.
Sodium butyrate protects against high-fat diet-induced oxidative stress in rat liver by promoting expression of nuclear factor E2-related factor 2.
The British journal of nutrition (2019-06-18)
Bo Sun, Yimin Jia, Shu Yang, Nannan Zhao, Yun Hu, Jian Hong, Shixing Gao, Ruqian Zhao
ABSTRACT
Oxidative stress is closely related to metabolic disorders, which can lead to various diseases. Nuclear factor E2-related factor 2 (Nrf2) is a central regulator of oxidative stress. Sodium butyrate (NaB) has been shown to alleviate oxidative stress and insulin resistance, yet how Nrf2 is involved in the action of NaB remains unclear. In the present study, rats were rendered obese by feeding a high-fat diet for 9 weeks. NaB (300 mg/kg), which was gavaged every 2 d for 7 weeks, significantly alleviated high-fat diet-induced oxidative stress and insulin resistance. Additionally, the insulin signalling pathway in the liver was activated by NaB, associated with significant activation of Nrf2, superoxide dismutase and glutathione. Furthermore, hepatic up-regulation of Nrf2 in NaB-treated rats was associated with reduced protein content of histone deacetylase 1 and increased histone H3 acetyl K9 (H3K9Ac) modification on the Nrf2 promoter. The actions of NaB were completely abolished when Nrf2 was knocked down in vitro. Taken together, NaB acts as a histone deacetylase inhibitor to up-regulate Nrf2 expression with enhanced H3K9Ac modification on its promoter. NaB-induced Nrf2 activation stimulates transcription of downstream antioxidant enzymes, thus contributing to the amelioration of high-fat diet-induced oxidative stress and insulin resistance.
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Ozagrel hydrochloride hydrate, ≥98% (HPLC), solid