  • Search Results
  • A Long Noncoding RNA, Antisense IL-7, Promotes Inflammatory Gene Transcription through Facilitating Histone Acetylation and Switch/Sucrose Nonfermentable Chromatin Remodeling.
A Long Noncoding RNA, Antisense IL-7, Promotes Inflammatory Gene Transcription through Facilitating Histone Acetylation and Switch/Sucrose Nonfermentable Chromatin Remodeling.
Journal of immunology (Baltimore, Md. : 1950) (2019-08-07)
Xu Liu, Yajing Lu, Jie Zhu, Mingjia Liu, Minghong Xie, Mengling Ye, Mingxuan Li, Shuhong Wang, Zhenping Ming, Qiang Tong, Feng Liu, Rui Zhou
ABSTRACT
Long noncoding RNAs are important regulators of gene expression in innate immune responses. Antisense IL-7 (IL-7-AS) is a newly discovered long noncoding RNA in human and mouse that has been reported to regulate the expression of IL-6. However, the potential function of IL-7-AS in innate immune system is not fully understood. In this study, we found that the expression of IL-7-AS is primarily dependent on the NF-κB and MAPK signaling pathways in macrophages and intestinal epithelial cells. Functionally, IL-7-AS promotes the expression of several inflammatory genes, including CCL2, CCL5, CCL7, and IL-6, in cells in response to LPS. Specifically, IL-7-AS physically interacts with p300 to regulate histone acetylation levels around the promoter regions of these gene loci. Moreover, IL-7-AS and p300 complex modulate the assembly of SWI/SNF complex to the promoters. IL-7-AS regulates chemotaxis activity of monocytes to intestine epithelial cells with involvement of CCL2. Therefore, our data indicate a new promoting role for NF-κB/MAPK-responsive IL-7-AS in the transcriptional regulation of inflammatory genes in the innate immune system although modulation of histone acetylation around the promoters of related genes.
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
C646, ≥98% (HPLC)