  • Search Results
  • A modified formulation of Chinese traditional medicine improves memory impairment and reduces Aβ level in the Tg-APPswe/PS1dE9 mouse model of Alzheimer's disease.
A modified formulation of Chinese traditional medicine improves memory impairment and reduces Aβ level in the Tg-APPswe/PS1dE9 mouse model of Alzheimer's disease.
Journal of ethnopharmacology (2011-07-19)
Songhee Jeon, Shambhunath Bose, Jinyoung Hur, Kiyoung Jun, Young-Kyoon Kim, Kyoung Sang Cho, Byung-Soo Koo
ABSTRACT
SuHeXiang Wan (SHXW), a Chinese traditional medicine has been used orally for the treatment of seizures, infantile convulsion, stroke and so forth. Previously, we reported the effects of modified SHXW essential oil mixture of the fragrance containing herbs on the sedative effect, anticonvulsant property and antioxidative activity after fragrance inhalation. This study was undertaken to evaluate beneficial effects of a modified recipe of SHXW (termed as KSOP1009) consisting of a ethanol extract of 8 herbs including resin of Liquidambar orientalis Miller, seed of Myristica fragrans Houtt., rhizome of Cnidium officinale Makino, lumber of Santalum album L., fructus of Piper longum L., flower buds of Eugenia caryophyllata Merrill et Perry, pollen of Typha orientalis Presl., and root of Salvia miltiorrhiza Bunge in the neurodegenerative diseases such as Alzheimer's disease (AD). The transgenic mice of AD, Tg-APPswe/PS1dE9, were fed KSOP1009 or as a positive control, donepezil for 3 months from 4.5 months of age. Behavioral, immunological and ELISA analyses were used to assess memory impairment, Aβ accumulation and plaque deposition in the brain. Other in vitro works were performed to examine whether KSOP1009 inhibits the Aβ(1-42)-induced neurotoxicity in human neuroblastoma cell line, SH-SY5Y cells. Intake of KSOP1009 improved the Aβ-induced memory impairment and suppressed Aβ levels and plaque deposition in the brain of Tg-APPswe/PS1dE9 mice as much as that of donepezil treatment. KSOP1009 prevented the down-regulation of phospho-CREB and increased AKT phosphorylation in the AD-like brains. Moreover, KSOP1009 suppresses Aβ-induced apoptosis and ROS production in SH-SY5Y cells. The present study suggests that KSOP1009 may develop as a therapeutic drug for treatment of AD patients.
Sigma-Aldrich
Monoclonal Anti-β-Amyloid antibody produced in mouse, clone BAM-10, ascites fluid
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.