  • Search Results
  • Cortical atrophy differentiates Richardson's syndrome from the parkinsonian form of progressive supranuclear palsy.
Cortical atrophy differentiates Richardson's syndrome from the parkinsonian form of progressive supranuclear palsy.
Movement disorders : official journal of the Movement Disorder Society (2011-03-18)
Emma C Schofield, John R Hodges, Virginia Macdonald, Nicholas J Cordato, Jillian J Kril, Glenda M Halliday
ABSTRACT
To determine whether brain atrophy differs between the two subtypes of progressive supranuclear palsy (PSP), Richardson's syndrome (PSP-RS), and PSP parkinsonism (PSP-P), and whether such atrophy directly relates to clinical deficits and the severity of tau deposition. We compared 24 pathologically confirmed PSP cases (17 PSP-RS and 7 PSP-P) with 22 controls from a Sydney brain donor program. Volume loss was analyzed in 29 anatomically discrete brain regions using a validated point-counting technique, and tau-immunoreactive neurons, astrocytes and oligodendrocytes/threads semiquantified. Correlations between the two pathological measures and the presence or absence of cardinal PSP symptoms were investigated. Cortical atrophy was more severe in PSP-RS than PSP-P and affected more frontal lobe regions (frontal pole, inferior frontal gyrus). The supramarginal gyrus was atrophic in both subtypes. Additionally, atrophy of the internal globus pallidus, amygdala, and thalamus was more severe in PSP-RS. As expected, more severe frontal lobe tau pathology differentiated PSP-RS from PSP-P. No correlations were found between the degree of atrophy and severity of tau pathology in any region assessed, or between the severity of atrophy or tau pathology and the presence or absence of cardinal PSP symptoms. Our study shows that thalamocortical atrophy is a defining feature of PSP-RS, but this atrophy does not correlate with the presence of any specific cardinal clinical feature. Interestingly, there is a disassociation between tau pathology and atrophy in the brain regions affected in PSP-RS that requires further investigation.
Sigma-Aldrich
Monoclonal Anti-τ (Tau) antibody produced in mouse, clone TAU-2, ascites fluid
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.