  • Search Results
  • Skeletal muscle regeneration is delayed by reduction in Xin expression: consequence of impaired satellite cell activation?
Skeletal muscle regeneration is delayed by reduction in Xin expression: consequence of impaired satellite cell activation?
American journal of physiology. Cell physiology (2011-10-07)
Aliyah A Nissar, Bart Zemanek, Rita Labatia, Daniel J Atkinson, Peter F M van der Ven, Dieter O Fürst, Thomas J Hawke
ABSTRACT
Xin is a striated muscle-specific actin-binding protein whose mRNA expression has been observed in damaged skeletal muscle. Here we demonstrate increased Xin protein expression early postinjury (≤ 12 h) and localization primarily to the periphery of damaged myofibers. At 1 day postinjury, Xin is colocalized with MyoD, confirming expression in activated satellite cells (SCs). By 5 days postinjury, Xin is evident in newly regenerated myofibers, with a return to preinjury levels by 14 days of regeneration. To determine whether the increased Xin expression is functionally relevant, tibialis anterior muscles of wild-type mice were infected with Xin-short hairpin RNA (shRNA) adenovirus, whereas the contralateral tibialis anterior received control adenovirus (Control). Four days postinfection, muscles were harvested or injured with cardiotoxin and collected at 3, 5, or 14 days thereafter. When compared with Control, Xin-shRNA infection attenuated muscle regeneration as demonstrated by Myh3 expression and fiber areas. Given the colocalization of Xin and MyoD, we isolated single myofibers from infected muscles to investigate the effect of silencing Xin on SC function. Relative to Control, SC activation, but not proliferation, was significantly impaired in Xin-shRNA-infected muscles. To determine whether Xin affects the G0-G1 transition, cell cycle reentry was assessed on infected C2C12 myoblasts using a methylcellulose assay. No difference in reentry was noted between groups, suggesting that Xin contributes to SC activation by means other than affecting G0-G1 transition. Together these data demonstrate a critical role for Xin in SC activation and reduction in Xin expression results in attenuated skeletal muscle repair.
Sigma-Aldrich
Monoclonal Anti-BrdU antibody produced in mouse, clone BU-33, ascites fluid, Immunohistology Grade
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.