  • Search Results
  • The N-terminal Ac-EEED sequence plays a role in alpha-smooth-muscle actin incorporation into stress fibers.
The N-terminal Ac-EEED sequence plays a role in alpha-smooth-muscle actin incorporation into stress fibers.
Journal of cell science (2005-03-17)
Sophie Clément, Boris Hinz, Vera Dugina, Giulio Gabbiani, Christine Chaponnier
ABSTRACT
We have previously shown that the N-terminal sequence AcEEED of alpha-smooth-muscle actin causes the loss of alpha-smooth-muscle actin from stress fibers and a decrease in cell contractility when introduced in myofibroblasts as a cell-penetrating fusion peptide. Here, we have investigated the function of this sequence on stress fiber organization in living cells, using enhanced green fluorescent protein (EGFP)-tagged alpha-smooth-muscle actin. The fusion peptide provokes the gradual disappearance of EGFP fluorescence of alpha-smooth-muscle actin from stress fibers and the formation of hitherto unknown rod-like structures. In addition to alpha-smooth-muscle actin, these structures contain cytoplasmic actins, gelsolin and cofilin but not other major actin-binding proteins. These rod-like structures are also visible in wild-type fibroblasts during normal cell spreading, suggesting that they represent a physiological step in the organization of alpha-smooth-muscle actin in stress fibers. Fluorescence-recovery-after-photobleaching experiments suggest that the fusion peptide reduces the dynamics of alpha-smooth-muscle actin and its incorporation in stress fibers. Here, we propose a new mechanism of how alpha-smooth-muscle actin is incorporated in stress fibers involving the sequence Ac-EEED.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Talin antibody produced in mouse, clone 8d4, ascites fluid