  • Search Results
  • Assessment of inter- and intra-specific variability in the main species of Boletus edulis complex by ITS analysis.
Assessment of inter- and intra-specific variability in the main species of Boletus edulis complex by ITS analysis.
FEMS microbiology letters (2005-02-03)
Marco Leonardi, Francesco Paolocci, Andrea Rubini, Giampaolo Simonini, Giovanni Pacioni
ABSTRACT
The Boletus edulis species complex includes ectomycorrhizal fungi producing edible mushrooms appreciated worldwide. However, species delineation is very difficult in these fungi, because it is based exclusively on a few, highly variable morphological features. As a consequence, a high number of taxa--including several varieties, subspecies and/or species sensu stricto--have been described in this species complex. In this paper we report on an extensive analysis of internal transcribed spacer of the nuclear rDNA region on a large sample of species of the B. edulis complex, mainly harvested in Italy, and representative of the European variability of this group. The molecular analysis allowed us to discriminate among and within B. edulis, B. aestivalis, B. pinophilus and B. aereus spp. and resolve their phylogenetic relationship.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.