  • Search Results
  • Different inactivating mutations in the LU genes of three individuals with the Lutheran-null phenotype.
Different inactivating mutations in the LU genes of three individuals with the Lutheran-null phenotype.
Transfusion (2007-02-27)
Vanja Karamatic Crew, Gary Mallinson, Carole Green, Joyce Poole, Makoto Uchikawa, Yoshihiko Tani, Christof Geisen, Johannes Oldenburg, Geoff Daniels
ABSTRACT
The null phenotype of the Lutheran blood group system, Lu(null) or Lu(a-b-), is characterized by the lack of all Lutheran system antigens. It can arise from three genetic backgrounds: recessive, dominant, or X-linked. Lu(null) of the recessive type appears to result from homozygosity for an inactive LU gene. Three unrelated recessive Lu(null) individuals were assessed by standard serologic tests. All exons of the LU gene were directly sequenced from amplified genomic DNA. The validity of the observed mutations within the LU gene was confirmed by the use of either restriction enzymes or allele-specific primers. All three individuals had the serologic characteristics of recessive Lu(null). One individual was doubly heterozygous for a nonsense mutation 691C>T in exon 6 (Arg231STOP) and a deletion of LU exons 3 and 4. The other two samples showed homozygous nonsense mutations: one had 711C>A in exon 6 (Cys237STOP) and the other 361C>T in exon 3 (Arg121STOP). The results revealed four unique genetic backgrounds from three examples of the rare recessive Lu(null) phenotype, each encoding Lutheran glycoproteins with a disrupted structure.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.