  • Search Results
  • Vinculin arrests motile B cells by stabilizing integrin clustering at the immune synapse.
Vinculin arrests motile B cells by stabilizing integrin clustering at the immune synapse.
Journal of immunology (Baltimore, Md. : 1950) (2013-07-23)
Julia Saez de Guinoa, Laura Barrio, Yolanda R Carrasco
ABSTRACT
Lymphocytes use integrin-based platforms to move and adhere firmly to the surface of other cells. The molecular mechanisms governing lymphocyte adhesion dynamics are however poorly understood. In this study, we show that in mouse B lymphocytes, the actin binding protein vinculin localizes to the ring-shaped integrin-rich domain of the immune synapse (IS); the assembly of this platform, triggered by cognate immune interactions, is needed for chemokine-mediated B cell motility arrest and leads to firm, long-lasting B cell adhesion to the APC. Vinculin is recruited early in IS formation, in parallel to a local phosphatidylinositol (4,5)-bisphosphate wave, and requires spleen tyrosine kinase activity. Lack of vinculin at the IS impairs firm adhesion, promoting, in turn, cell migration with Ag clustered at the uropod. Vinculin localization to the B cell contact area depends on actomyosin. These results identify vinculin as a major controller of integrin-mediated adhesion dynamics in B cells.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Talin antibody produced in mouse, clone 8d4, ascites fluid