  • Search Results
  • Identification of B Cell Epitopes of Blo t 13 Allergen and Cross-Reactivity with Human Adipocytes and Heart Fatty Acid Binding Proteins.
Identification of B Cell Epitopes of Blo t 13 Allergen and Cross-Reactivity with Human Adipocytes and Heart Fatty Acid Binding Proteins.
International journal of molecular sciences (2019-12-11)
Marlon Múnera, Dalgys Martínez, Alexis Labrada, Luis Caraballo, Leonardo Puerta
ABSTRACT
Cross-reactivity between allergens and human proteins could have a clinical impact in allergic diseases. Blo t 13 is an allergen from the mite Blomia tropicalis, which belongs to the fatty acid binding protein (FABP) family and has structural homology with human FABPs. This work aimed to map B cell epitopes on Blo t 13 and to identify epitopes involved in cross-reactivity with human heart FABP (FABP3) and adipocyte FABP (FABP4). Sera from 25 patients with house dust mite (HDM) allergy that were sensitized to Blo t 13 were used for testing the reactivity of immunoglobulin E (IgE) and IgG to FABP. The epitope mapping of Blo t 13 was performed using overlapping peptides, and cross-reactivity between Blo t 13 and human FABP was analyzed using human sera and anti-Blo t 13 monoclonal antibodies. IgE antibodies to all FABPs were detected in 14/25 serum samples, and IgG was detected in 25/25 serum samples. The cross-reactivity of Blo t 13 was 42% with FABP3 and 48% with FABP4. Two IgE-binding regions were identified in Blo t 13; one between residues 54 and 72 (the main cross-reacting region) and another between residues 111 to 129. Our results suggest that exposure to the Blo t 13 allergen could induce an auto-reactive response to endogenous FABP in allergic patients sensitized to Blo t 13.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid
Sigma-Aldrich
Anti-Human IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgE (ε-chain specific)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.