PCR-RFLP analysis of mitochondrial DNA in tench Tinca tinca.
Journal of fish biology (2010-08-27)
R Lo Presti, L Gasco, C Lisa, I Zoccarato, L Di Stasio
ABSTRACT
Polymorphism was detected at ND1, ND6, D-loop and cyt b segments of mtDNA in 105 tench (Tinca tinca L.), using the polymerase chain reaction-restriction fragment length polymorphism (PCR-RFLP) technique and five composite haplotypes were identified. The diversity indices and the results of the population comparisons revealed that the identified markers provide a powerful tool for further studies on this species.
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.