  • Search Results
  • Anti-inflammatory Role of Carotenoids in Endothelial Cells Derived from Umbilical Cord of Women Affected by Gestational Diabetes Mellitus.
Anti-inflammatory Role of Carotenoids in Endothelial Cells Derived from Umbilical Cord of Women Affected by Gestational Diabetes Mellitus.
Oxidative medicine and cellular longevity (2019-03-29)
Mariangela Ucci, Pamela Di Tomo, Federica Tritschler, Vincenzo G P Cordone, Paola Lanuti, Giuseppina Bologna, Sara Di Silvestre, Natalia Di Pietro, Caterina Pipino, Domitilla Mandatori, Gloria Formoso, Assunta Pandolfi
ABSTRACT
Diabetes is associated with vascular inflammation, endothelial dysfunction, and oxidative stress, promoting the development of cardiovascular diseases (CVD). Several studies showed that a carotenoid-rich diet is associated to a reduced cardiovascular risk in healthy and diabetic subjects, although the mechanisms of action are still unknown. Here, the potential role of β-carotene (BC) and lycopene (Lyc) in human endothelial cells isolated from human umbilical cord vein (HUVECs) of women with gestational diabetes (GD) and respective controls (C) has been investigated. Results showed that BC and Lyc reduced the tumor necrosis factor alpha- (TNF-α-) stimulated monocyte-endothelium interaction (adhesion assay), membrane exposure (flow cytometry), and total expression levels (Western blot) of VCAM-1 and ICAM-1 in both cell types. Moreover, the treatment with BC and Lyc reduced the TNF-α-induced nuclear translocation of NF-κB (image flow cytometry) by preserving bioavailability of nitric oxide (NO, flow cytometry, and cGMP EIA kit assay), a key vasoactive molecule. Notably, BC and Lyc pretreatment significantly reduced peroxynitrite levels (flow cytometry), contributing to the redox balance protection. These results suggest a new mechanism of action of carotenoids which exert vascular protective action in diabetic condition, thus reinforcing the importance of a carotenoid-rich diet in the prevention of diabetes cardiovascular complications.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
SAFC
Trypsin-EDTA Solution 10X