  • Search Results
  • A Novel Anti-EGFR mAb Ame55 with Lower Toxicity and Better Efficacy than Cetuximab When Combined with Irinotecan.
A Novel Anti-EGFR mAb Ame55 with Lower Toxicity and Better Efficacy than Cetuximab When Combined with Irinotecan.
Journal of immunology research (2019-02-09)
Weiyi Qiu, Chang Zhang, Shuang Wang, Xiaoyan Yu, Qiong Wang, Dadi Zeng, Peng Du, Jinling Ma, Yiqiong Zheng, Bo Pang, Yunzhou Yu, Feng Long, Xiaobin Pang, Zhiwei Sun
ABSTRACT
To improve efficacy and minimize toxicity of EGFR inhibition treatment, we developed Ame55, a novel anti-EGFR IgG1 with lower affinity to EGFR than cetuximab (C225) from a human phage library. Ame55 had lower bioactivity than cetuximab in vitro but similar antitumor efficacy as cetuximab in vivo. Moreover, Ame55 was more efficacious than cetuximab in a Lovo cell xenograft tumor model when combined with irinotecan (CPT-11). Ame55 concentrates in the mouse xenograft tumor and has less toxicity than cetuximab in cynomolgus monkeys in an overdose study.
Sigma-Aldrich
Anti-Human IgG (Fc specific)−Peroxidase antibody produced in goat, affinity isolated antibody
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.