  • Search Results
  • Hydroxamic acids as a novel family of serine racemase inhibitors: mechanistic analysis reveals different modes of interaction with the pyridoxal-5'-phosphate cofactor.
Hydroxamic acids as a novel family of serine racemase inhibitors: mechanistic analysis reveals different modes of interaction with the pyridoxal-5'-phosphate cofactor.
Journal of medicinal chemistry (2009-10-02)
Hillary E Hoffman, Jana Jirásková, Petr Cígler, Miloslav Sanda, Jan Schraml, Jan Konvalinka
ABSTRACT
Mammalian serine racemase (SR) is a pyridoxal-5'-phosphate (PLP) dependent enzyme responsible for the biosynthesis of the neurotransmitter D-serine, which activates N-methyl-D-aspartate (NMDA) receptors in the CNS. Aberrant regulation of NMDA receptor signaling has been implicated in a variety of neuropathologies, and inhibitors of SR would therefore be a worthwhile tool for further investigation or treatment of such conditions. Here, we identify a series of small aliphatic hydroxamic acids (HAs) that act as potent SR inhibitors. However, specificity studies showed that some of these HAs can act as nonspecific inhibitors of PLP-dependent enzymes. We employed NMR, MS, and UV/vis spectroscopic techniques to reveal that the nonspecific effect is likely due to irreversible interaction of the HA moiety with PLP to form aldoxime species. We also characterize L-aspartic acid beta-hydroxamate as a competitive and selective SR inhibitor that could be used as a scaffold for further inhibitor development.
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Serine, certified reference material, TraceCERT®
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Aspartic acid β-hydroxamate, serine racemase inhibitor
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
SAFC
Glycine
Sigma-Aldrich
Acetohydroxamic acid, 98%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Malonic acid, ReagentPlus®, 99%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Acetaldehyde oxime, mixture of syn and anti, 99%