  • Search Results
  • Pharmacologic and genetic approaches define human pancreatic β cell mitogenic targets of DYRK1A inhibitors.
Pharmacologic and genetic approaches define human pancreatic β cell mitogenic targets of DYRK1A inhibitors.
JCI insight (2019-12-11)
Courtney Ackeifi, Ethan Swartz, Kunal Kumar, Hongtao Liu, Suebsuwong Chalada, Esra Karakose, Donald K Scott, Adolfo Garcia-Ocaña, Roberto Sanchez, Robert J DeVita, Andrew F Stewart, Peng Wang
ABSTRACT
Small molecule inhibitors of dual specificity, tyrosine phosphorylation-regulated kinase 1A (DYRK1A), including harmine and others, are able to drive human β cell regeneration. While DYRK1A is certainly a target of this class, whether it is the only or the most important target is uncertain. Here, we employ a combined pharmacologic and genetic approach to refine the potential mitogenic targets of the DYRK1A inhibitor family in human islets. A combination of human β cell RNA sequencing, DYRK1A inhibitor kinome screens, pharmacologic inhibitors, and targeted silencing of candidate genes confirms that DYRK1A is a central target. Surprisingly, however, DYRK1B also proves to be an important target: silencing DYRK1A results in an increase in DYRK1B. Simultaneous silencing of both DYRK1A and DYRK1B yields greater β cell proliferation than silencing either individually. Importantly, other potential kinases, such as the CLK and the GSK3 families, are excluded as important harmine targets. Finally, we describe adenoviruses that are able to silence up to 7 targets simultaneously. Collectively, we report that inhibition of both DYRK1A and DYRK1B is required for induction of maximal rates of human β cell proliferation, and we provide clarity for future efforts in structure-based drug design for human β cell regenerative drugs.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Harmine, 98%
Sigma-Aldrich
TG003, ≥98% (HPLC)
Sigma-Aldrich
Anti-DYRK1A (N-terminal) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
AZ191, ≥98% (HPLC)