MilliporeSigma
  • Evolution of carnivorous traps from planar leaves through simple shifts in gene expression.
Evolution of carnivorous traps from planar leaves through simple shifts in gene expression.
Science (New York, N.Y.) (2019-11-23)
Christopher D Whitewoods, Beatriz Gonçalves, Jie Cheng, Minlong Cui, Richard Kennaway, Karen Lee, Claire Bushell, Man Yu, Chunlan Piao, Enrico Coen
ABSTRACT
Leaves vary from planar sheets and needle-like structures to elaborate cup-shaped traps. Here, we show that in the carnivorous plant Utricularia gibba, the upper leaf (adaxial) domain is restricted to a small region of the primordium that gives rise to the trap's inner layer. This restriction is necessary for trap formation, because ectopic adaxial activity at early stages gives radialized leaves and no traps. We present a model that accounts for the formation of both planar and nonplanar leaves through adaxial-abaxial domains of gene activity establishing a polarity field that orients growth. In combination with an orthogonal proximodistal polarity field, this system can generate diverse leaf forms and account for the multiple evolutionary origins of cup-shaped leaves through simple shifts in gene expression.
Sigma-Aldrich
1-Naphthaleneacetic acid, suitable for plant cell culture, BioReagent, ≥95%, crystalline
Sigma-Aldrich
6-Benzylaminopurine, suitable for plant cell culture
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Cefotaxime sodium salt, suitable for plant cell culture, BioReagent, powder or crystals
Sigma-Aldrich
3′,5′-Dimethoxy-4′-hydroxyacetophenone, 97%