  • Search Results
  • Hypertonic external medium represses cellular respiration and promotes Warburg/Crabtree effect.
Hypertonic external medium represses cellular respiration and promotes Warburg/Crabtree effect.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-01-10)
Minoo Hamraz, Raymond Abolhassani, Mireille Andriamihaja, Céline Ransy, Véronique Lenoir, Laurent Schwartz, Frédéric Bouillaud
ABSTRACT
Hyperosmotic conditions are associated to several pathological states. In this article, we evaluate the consequence of hyperosmotic medium on cellular energy metabolism. We demonstrate that exposure of cells to hyperosmotic conditions immediately reduces the mitochondrial oxidative phosphorylation rate. This causes an increase in glycolysis, which represses further respiration. This is known as the Warburg or Crabtree effect. In addition to aerobic glycolysis, we observed two other cellular responses that would help to preserve cellular ATP level and viability: A reduction in the cellular ATP turnover rate and a partial mitochondrial uncoupling which is expected to enhance ATP production by Krebs cycle. The latter is likely to constitute another metabolic adaptation to compensate for deficient oxidative phosphorylation that, importantly, is not dependent on glucose.
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, BioXtra, ≥99% (HPLC), from microbial
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Potassium cyanide, BioUltra, ≥98.0% (AT)
Sigma-Aldrich
L-Glutamic acid monosodium salt hydrate, ≥99% (HPLC), powder
Sigma-Aldrich
Digitonin, ~50% (TLC)
Sigma-Aldrich
Poly(ethylene glycol), average mol wt 400
Sigma-Aldrich
L-(−)-Malic acid, suitable for cell culture, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Adenosine 5′-diphosphate bis(cyclohexylammonium) salt, ≥95% (HPLC), crystalline, bacterial
Sigma-Aldrich
Monoclonal Anti-S100A6 antibody produced in mouse, clone CACY-100, ascites fluid