  • Search Results
  • Enhanced differentiation of human dopaminergic neuronal cell model for preclinical translational research in Parkinson's disease.
Enhanced differentiation of human dopaminergic neuronal cell model for preclinical translational research in Parkinson's disease.
Biochimica et biophysica acta. Molecular basis of disease (2019-08-24)
Dilshan S Harischandra, Dharmin Rokad, Shivani Ghaisas, Saurabh Verma, Alan Robertson, Huajun Jin, Vellareddy Anantharam, Arthi Kanthasamy, Anumantha G Kanthasamy
ABSTRACT
Human-derived neuronal cell lines are progressively being utilized in understanding neurobiology and preclinical translational research as they are biologically more relevant than rodent-derived cells lines. The Lund human mesencephalic (LUHMES) cell line comprises human neuronal cells that can be differentiated to post-mitotic neurons and is increasingly being used as an in vitro model for various neurodegenerative diseases. A previously published 2-step differentiation procedure leads to the generation of post-mitotic neurons within 5-days, but only a small proportion (10%) of the total cell population tests positive for tyrosine hydroxylase (TH). Here we report on a novel differentiation protocol that we optimized by using a cocktail of neurotrophic factors, pleiotropic cytokines, and antioxidants to effectively generate proportionately more dopaminergic neurons within the same time period. Visualization and quantification of TH-positive cells revealed that under our new protocol, 25% of the total cell population expressed markers of dopaminergic neurons with the TH-positive neuron count peaking on day 5. These neurons showed spontaneous electrical activity and responded to known Parkinsonian toxins as expected by showing decreased cell viability and dopamine uptake and a concomitant increase in apoptotic cell death. Together, our results outline an improved method for generating a higher proportion of dopaminergic neurons, thus making these cells an ideal neuronal culture model of Parkinson's disease (PD) for translational research.
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
Anti-Dopamine Transporter Antibody, NT, clone DAT-Nt, culture supernatant, clone DAT-Nt, Chemicon®
Sigma-Aldrich
Atto Rho12
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.