  • Search Results
  • Renewable Responsive Systems Based on Original Click and Polyurethane Cross-Linked Architectures with Advanced Properties.
Renewable Responsive Systems Based on Original Click and Polyurethane Cross-Linked Architectures with Advanced Properties.
ChemSusChem (2019-09-07)
Khantutta-Kim Tremblay-Parrado, Luc Avérous
ABSTRACT
A new chemical architecture from oleic acid, consisting of a diol structure containing pendant furan rings, denoted the furan oligomer (FO) was synthesized and fully characterized. The FO was integrated into a linear rapeseed-based polyurethane (PU) backbone and cross-linked through a Diels-Alder (DA) reaction by using pendant furan rings and a short polypropylene oxide-based bismaleimide. This is the first time that a thermoreversible PU network based on vegetable oil has been reported. The effects of varying proportions of FO in linear and cross-linked systems, by DA, were studied. These materials were analyzed by classic characterization techniques. The stability and recyclability of the cross-linked materials were shown by successive reprocessing cycles and reanalyzing the mechanical properties. Self-healing properties were macroscopically exhibited and investigated by tensile tests on healed materials. The resulting cross-linked materials present a large range of properties, such as tunable mechanical and thermoresponsive behavior, good thermal recyclability, and self-healing abilities.
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
2,3,4,5,6-Pentafluorobenzaldehyde, 98%
Sigma-Aldrich
2,4-Dihydroxybenzophenone, 99%
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.