  • Search Results
  • Transcription factor ZFHX3 regulates calcium influx in mammary epithelial cells in part via the TRPV6 calcium channel.
Transcription factor ZFHX3 regulates calcium influx in mammary epithelial cells in part via the TRPV6 calcium channel.
Biochemical and biophysical research communications (2019-09-15)
Dan Zhao, Xueying Han, Lili Huang, Jianpeng Wang, Xi Zhang, Ju-Hong Jeon, Qiang Zhao, Jin-Tang Dong
ABSTRACT
Zinc finger homeobox 3 (ZFHX3) is a transcription factor that regulates multiple cellular processes including cell proliferation, differentiation and neoplastic development. It is also involved in the function of steroid hormones estrogen and progesterone and the peptide hormone prolactin in mammary epithelial cells. In this study, we investigated whether and how ZFHX3 regulates intracellular calcium homeostasis in mammary epithelial cells. We found that ZFHX3 affected both store operated calcium entry and store independent calcium entry (SOCE and SICE). Simultaneously, the expression of the calcium channel TRPV6 was regulated by ZFHX3, as demonstrated by expression analysis and luciferase reporter assay. In cells with knockdown of ZFHX3, calcium entry was partially rescued by the overexpression of wild type but not the pore mutants of TRPV6. In addition, overexpression of TRPV6 promoted differentiation of the MCF10A mammary epithelial cells in three-dimensional culture, which is consistent with our previous findings that ZFHX3 is essential for mammary gland differentiation. These findings suggest that ZFHX3 plays an important role in intracellular calcium homeostasis in mammary epithelial cells, at least in part, by regulating TRPV6.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Fura 2-AM, ≥95% (HPLC)