  • Search Results
  • Assessment of Enzymatic Improvers in Flours Using LC-MS/MS Detection of Marker Tryptic Peptides.
Assessment of Enzymatic Improvers in Flours Using LC-MS/MS Detection of Marker Tryptic Peptides.
Journal of the American Society for Mass Spectrometry (2020-02-08)
Barbara Prandi, Francesca Lambertini, Martina Varani, Andrea Faccini, Michele Suman, Andrea Leporati, Tullia Tedeschi, Stefano Sforza
ABSTRACT
Enzymatic improvers are enzymes obtained from microbial or fungal cultures, added as technical adjuvants to flour, with the aim of improving the dough characteristics in bakery products. They are used in a low ppm range and, being technical adjuvants, can go undeclared on the label. Many types of enzymatic improvers are present on the market, such as amylases, lipases, proteases, xylanases, glucose oxidases, and others, each with a different function. Analytical methods capable of detecting these enzymes are needed, particularly for bakery companies, in order to monitor the quality of raw materials and to detect any undeclared presence. In the present work, specific peptide markers, obtained by enzymatic digestion, have been used to detect the presence of enzymatic improvers by LC-MS/MS techniques. Promising results were obtained for some enzymes acting on the carbohydrate fraction (glucoamylase, glucose oxidase, xylanase) in which amounts as low as 20 ppm could be identified in blind flour samples. For lipases and proteases the method proved to be very effective in terms of specific identification, even if less sensitive.
Sigma-Aldrich
Calcium chloride, anhydrous, granular, ≤7.0 mm, ≥93.0%
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%