  • Search Results
  • An integrated perfusion machine preserves injured human livers for 1 week.
An integrated perfusion machine preserves injured human livers for 1 week.
Nature biotechnology (2020-01-15)
Dilmurodjon Eshmuminov, Dustin Becker, Lucia Bautista Borrego, Max Hefti, Martin J Schuler, Catherine Hagedorn, Xavier Muller, Matteo Mueller, Christopher Onder, Rolf Graf, Achim Weber, Philipp Dutkowski, Philipp Rudolf von Rohr, Pierre-Alain Clavien
ABSTRACT
The ability to preserve metabolically active livers ex vivo for 1 week or more could allow repair of poor-quality livers that would otherwise be declined for transplantation. Current approaches for normothermic perfusion can preserve human livers for only 24 h. Here we report a liver perfusion machine that integrates multiple core physiological functions, including automated management of glucose levels and oxygenation, waste-product removal and hematocrit control. We developed the machine in a stepwise fashion using pig livers. Study of multiple ex vivo parameters and early phase reperfusion in vivo demonstrated the viability of pig livers perfused for 1 week without the need for additional blood products or perfusate exchange. We tested the approach on ten injured human livers that had been declined for transplantation by all European centers. After a 7-d perfusion, six of the human livers showed preserved function as indicated by bile production, synthesis of coagulation factors, maintained cellular energy (ATP) and intact liver structure.
Sigma-Aldrich
Drabkin′s reagent, for the determination of hemoglobin
Sigma-Aldrich
Glycogen Assay Kit, sufficient for 100 colorimetric or fluorometric tests