  • Search Results
  • Silencing of Long Non-Coding RNA Colon Cancer-Associated Transcript 2 Inhibits the Growth and Metastasis of Gastric Cancer Through Blocking mTOR Signaling.
Silencing of Long Non-Coding RNA Colon Cancer-Associated Transcript 2 Inhibits the Growth and Metastasis of Gastric Cancer Through Blocking mTOR Signaling.
OncoTargets and therapy (2020-02-06)
Sen Lin, Hongbo Wang, Wenjuan Yang, Aiguang Wang, Chao Geng
ABSTRACT
This study aimed to evaluate the specific role of colon cancer-associated transcript 2 (CCAT2) on gastric cancer (GC), and reveal the potential regulatory mechanism relating to mammalian target of rapamycin (mTOR) signaling. The expression of CCAT2 was detected in GC tissues and cells by quantitative real-time PCR (qRT-PCR), and its relation with the pathologic characteristics of GC patients was analyzed. HGC-27 and SGC-7901 cells were transfected with siRNA-CCAT2 to silence CCAT2, and HGC-27 cells were then treated with an mTOR agonist Leucine (Leu) to activate mTOR signaling. The cell proliferation was evaluated by cell viability and colony formation. The cell cycle and apoptosis, and the migration and invasion abilities were detected by Flow cytometry, and Transwell assay, respectively. The expression of PCNA (proliferation marker), Snail, N-cadherin, E-cadherin (invasion markers), P53, Caspase-8, Bcl-2 (apoptosis markers), LC3-II/LC3-I, ATG3, p62 (autophagy makers), phosphorylated mTOR (p-mTOR), p-AKT, and p-p70S6K (mTOR signaling markers) were detected by Western blot. CCAT2 was upregulated in GC tissues and cells, and positively associated with the maximum tumor diameter, lymphatic metastasis, TNM staging, and low overall survival rate (P < 0.05). siRNA-CCAT2 transfection significantly inhibited the viability, colony formation, and migration and invasion abilities, blocked the cell cycle in G0/G1 phase, and promoted the apoptosis and autophagy of SGC-7901 and HGC-27 cells (P < 0.05). In addition, siRNA-CCAT2 transfection significantly upregulated P53, Caspase-8, LC3-II/LC3-I and ATG3, and downregulated PCNA, Bcl-2, p62, p-mTOR, p-AKT and p-p70S6K in SGC-7901 and HGC-27 cells (P < 0.05). siRNA-CCAT2 reversed the tumor-promoting effect of mTOR signaling activation on HGC-27 cells (P < 0.05). Silencing of CCAT2 inhibited the proliferation, migration and invasion, and promoted the apoptosis and autophagy of GC cells through blocking mTOR signaling.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1