MilliporeSigma
  • Measuring mRNA translation in neuronal processes and somata by tRNA-FRET.
Measuring mRNA translation in neuronal processes and somata by tRNA-FRET.
Nucleic acids research (2020-01-25)
Bella Koltun, Sivan Ironi, Noga Gershoni-Emek, Iliana Barrera, Mohammad Hleihil, Siddharth Nanguneri, Ranjan Sasmal, Sarit S Agasti, Deepak Nair, Kobi Rosenblum
ABSTRACT
In neurons, the specific spatial and temporal localization of protein synthesis is of great importance for function and survival. Here, we visualized tRNA and protein synthesis events in fixed and live mouse primary cortical culture using fluorescently-labeled tRNAs. We were able to characterize the distribution and transport of tRNAs in different neuronal sub-compartments and to study their association with the ribosome. We found that tRNA mobility in neural processes is lower than in somata and corresponds to patterns of slow transport mechanisms, and that larger tRNA puncta co-localize with translational machinery components and are likely the functional fraction. Furthermore, chemical induction of long-term potentiation (LTP) in culture revealed up-regulation of mRNA translation with a similar effect in dendrites and somata, which appeared to be GluR-dependent 6 h post-activation. Importantly, measurement of protein synthesis in neurons with high resolutions offers new insights into neuronal function in health and disease states.
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-Tubulin α antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Puromycin Antibody, clone 4G11, clone 4G11, from mouse
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, purified from hybridoma cell culture
Sigma-Aldrich
Poly-L-ornithine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture