  • Search Results
  • PPAR-gamma agonist Azelaoyl PAF increases frataxin protein and mRNA expression: new implications for the Friedreich's ataxia therapy.
PPAR-gamma agonist Azelaoyl PAF increases frataxin protein and mRNA expression: new implications for the Friedreich's ataxia therapy.
Cerebellum (London, England) (2008-12-24)
Daniele Marmolino, Fabio Acquaviva, Michele Pinelli, Antonella Monticelli, Imma Castaldo, Alessandro Filla, Sergio Cocozza
ABSTRACT
Friedreich's ataxia is a neurodegenerative disease due to frataxin deficiency, and thus, drugs increasing the frataxin amount are excellent candidates for therapy. By screening Gene Expression Omnibus profiles, we identified records showing a frataxin response to the peroxisome proliferator-activated receptors gamma (PPAR-gamma) agonist rosiglitazone. We decided to investigate the effect of the PPAR-gamma agonist Azelaoyl PAF on the frataxin protein and mRNA expression profile. We treated human neuroblastoma cells SKNBE and primary fibroblasts from skin biopsies from Friedreich's ataxia (FRDA) patients and healthy controls with the PPAR-gamma agonist Azelaoyl PAF. We show in this paper for the first time that Azelaoyl PAF significantly increases the intracellular frataxin levels by twofold in the neuroblastoma cell line SKNBE and fibroblasts from FRDA patients and that Azelaoyl PAF increases frataxin protein through a transcriptional mechanism. PPAR-gamma agonist Azelaoyl PAF increases both messenger RNA and protein levels of frataxin. We hypothesize that PPAR-gamma agonists could play a role in the treatment of FRDA, and our results offer the logical bases to further investigate the usefulness of this group of agents for the treatment of the FRDA.
Sigma-Aldrich
Azelaoyl PAF, 10 mg/mL in ethanol
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.