  • Search Results
  • Ordered structure-forming properties of the intrinsically disordered AB region of hRXRγ and its ability to promote liquid-liquid phase separation.
Ordered structure-forming properties of the intrinsically disordered AB region of hRXRγ and its ability to promote liquid-liquid phase separation.
The Journal of steroid biochemistry and molecular biology (2019-12-28)
Katarzyna Sołtys, Andrzej Ożyhar
ABSTRACT
The retinoid X receptor (RXR) is a member of the nuclear receptor (NR) superfamily that occupies the central position among other NRs by forming both homodimers and heterodimers with other representatives of the family. RXR shares similar structural domains with other members of NRs. The major differences in the subtypes and isoforms of RXR are in the AB region. To date, there have been no data concerning the molecular properties of the AB region of hRXRγ (AB_hRXG). Here, we describe the biochemical and biophysical properties of the recombinant AB_hRXG. The results indicate that AB_hRXG shows the structural and functional characteristics of the pre-molten globule-like (PMG-like) group of intrinsically disordered proteins (IDPs) and also has a significant propensity for folding. We also present the first experimental evidence showing that the AB region of NRs promotes the formation of liquid-liquid phase separation (LLPS).
Sigma-Aldrich
Atto 488 NHS ester, BioReagent, suitable for fluorescence, ≥80% (coupling to amines)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.