  • Search Results
  • FXR1 splicing is important for muscle development and biomolecular condensates in muscle cells.
FXR1 splicing is important for muscle development and biomolecular condensates in muscle cells.
The Journal of cell biology (2020-04-25)
Jean A Smith, Ennessa G Curry, R Eric Blue, Christine Roden, Samantha E R Dundon, Anthony Rodríguez-Vargas, Danielle C Jordan, Xiaomin Chen, Shawn M Lyons, John Crutchley, Paul Anderson, Marko E Horb, Amy S Gladfelter, Jimena Giudice
ABSTRACT
Fragile-X mental retardation autosomal homologue-1 (FXR1) is a muscle-enriched RNA-binding protein. FXR1 depletion is perinatally lethal in mice, Xenopus, and zebrafish; however, the mechanisms driving these phenotypes remain unclear. The FXR1 gene undergoes alternative splicing, producing multiple protein isoforms and mis-splicing has been implicated in disease. Furthermore, mutations that cause frameshifts in muscle-specific isoforms result in congenital multi-minicore myopathy. We observed that FXR1 alternative splicing is pronounced in the serine- and arginine-rich intrinsically disordered domain; these domains are known to promote biomolecular condensation. Here, we show that tissue-specific splicing of fxr1 is required for Xenopus development and alters the disordered domain of FXR1. FXR1 isoforms vary in the formation of RNA-dependent biomolecular condensates in cells and in vitro. This work shows that regulation of tissue-specific splicing can influence FXR1 condensates in muscle development and how mis-splicing promotes disease.
Sigma-Aldrich
Atto 488, suitable for fluorescence, ≥90% (HPCE)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.