  • Search Results
  • Comparative study on the plasma lipid oxidation induced by peroxynitrite and peroxyl radicals and its inhibition by antioxidants.
Comparative study on the plasma lipid oxidation induced by peroxynitrite and peroxyl radicals and its inhibition by antioxidants.
Free radical research (2019-11-07)
Mayuko Morita, Yuji Naito, Yoshito Itoh, Etsuo Niki
ABSTRACT
The unregulated oxidative modification of biological molecules has been implicated in the pathogenesis of various diseases, and the beneficial effects of antioxidants against detrimental oxidation have received much attention. Among the multiple oxidants, peroxyl radical and peroxynitrite play an important role as chain-carrying species in lipid peroxidation and one of the major oxidants produced in vivo, respectively. This study was performed to elucidate the prominent features of these two oxidants by comparing their reactivity and selectivity and also the effects of antioxidants against plasma lipid oxidation induced by the two oxidants. It was shown that despite peroxyl radical and peroxynitrite gave similar pattern of lipid peroxidation products of plasma, and these two oxidants exert different selectivity and reactivity towards probes and antioxidants. The capacity of antioxidants to scavenge peroxynitrite and peroxyl radical decreased in the order BSA > glutathione > α-tocopherol ∼ bilirubin ∼ α - tocotrienol > γ-tocotrienol ∼ γ - tocopherol > uric acid and α-tocopherol ∼ α - tocotrienol > bilirubin > γ-tocotrienol ∼ γ - tocopherol > BSA > glutathione > uric acid, respectively. α-Tocopherol localised within plasma lipoproteins was six times less effective than trolox in aqueous phase for scavenging peroxynitrite and the derived oxidants, despite the same chemical reactivity of the two chromanols. BSA was relatively more effective as antioxidant against peroxynitrite than peroxyl radical, whereas TEMPO did not act as efficient antioxidant against both oxidants. It was suggested that thiols act as more potent antioxidant against peroxynitrite than phenolic antioxidants, while phenolic antioxidants are potent inhibitor of lipid peroxidation induced by free radicals including those derived from peroxynitrite. Abbreviations: AAPH: 2,2'-azobis(2-amidinopropane) dihydrochloride; C11-BODIPY: 4,4-difluoro-5-(4-phenyl-1,3-butadienyl)-4-bora-3a,4a-diaza-s-indacene-3-undecanoic acid; BSA: bovine serum albumin; DPPP: diphenyl-1-pyrenylphosphine; H(p)ODE: hydro(pero)xyoctadecadienoates; PGR: pyrogallol red; PUFA: polyunsaturated fatty acid; SIN-1: 3-morpholinosydnonimine; TEMPO: 2,2-6,6 tetramethylpiperidine-1-oxyl; Trolox: 2-carboxy-2,5,7,8-tetramethyl-6-hydroxychroman.
Sigma-Aldrich
8-Hydroxypyrene-1,3,6-trisulfonic acid trisodium salt, ≥96%
Sigma-Aldrich
Pyrogallol Red
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile) solution, 0.2 M in toluene
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%