  • Search Results
  • Forkhead Box M1 Transcription Factor Drives Liver Inflammation Linking to Hepatocarcinogenesis in Mice.
Forkhead Box M1 Transcription Factor Drives Liver Inflammation Linking to Hepatocarcinogenesis in Mice.
Cellular and molecular gastroenterology and hepatology (2019-11-02)
Tomohide Kurahashi, Yuichi Yoshida, Satoshi Ogura, Mayumi Egawa, Kunimaro Furuta, Hayato Hikita, Takahiro Kodama, Ryotaro Sakamori, Shinichi Kiso, Yoshihiro Kamada, I-Ching Wang, Hidetoshi Eguchi, Eiichi Morii, Yuichiro Doki, Masaki Mori, Vladimir V Kalinichenko, Tomohide Tatsumi, Tetsuo Takehara
ABSTRACT
Liver inflammation has been recognized as a hallmark of hepatocarcinogenesis. Although Forkhead Box M1 (FoxM1) is a well-defined oncogenic transcription factor that is overexpressed in hepatocellular carcinoma (HCC), its role in liver inflammation has never been explored. We generated hepatocyte-specific FoxM1 conditional transgenic (TG) mice by using the Cre-loxP and Tetracycline (Tet)-on systems to induce FoxM1 expression in a hepatocyte-specific and time-dependent manner. After treatment of Tet-derivatives doxycycline (DOX) to induce FoxM1, TG mice exhibited spontaneous development of hepatocyte death with elevated serum alanine aminotransferase levels and hepatic infiltration of macrophages. The removal of DOX in TG mice completely removed this effect, suggesting that spontaneous inflammation in TG mice occurs in a hepatocyte FoxM1-dependent manner. In addition, liver inflammation in TG mice was associated with increased levels of hepatic and serum chemokine (C-C motif) ligand 2 (CCL2). In vitro transcriptional analysis confirmed that CCL2 is a direct target of FoxM1 in murine hepatocytes. After receiving FoxM1 induction since birth, all TG mice exhibited spontaneous HCC with liver fibrosis at 12 months of age. Hepatic expression of FoxM1 was significantly increased in liver injury models. Finally, pharmacologic inhibition of FoxM1 reduced liver inflammation in models of liver injury. Hepatocyte FoxM1 acts as a crucial regulator to orchestrate liver inflammation linking to hepatocarcinogenesis. Thus, hepatocyte FoxM1 may be a potential target not only for the treatment of liver injury but also for the prevention toward HCC.
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Thiostrepton from Streptomyces azureus, ≥90% (HPLC)
Sigma-Aldrich
ApopTag Peroxidase In Situ Apoptosis Detection Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.