  • Search Results
  • IL-33 Exacerbates Endometriotic Lesions via Polarizing Peritoneal Macrophages to M2 Subtype.
IL-33 Exacerbates Endometriotic Lesions via Polarizing Peritoneal Macrophages to M2 Subtype.
Reproductive sciences (Thousand Oaks, Calif.) (2020-02-13)
Yosuke Ono, Osamu Yoshino, Takehiro Hiraoka, Ikumi Akiyama, Erina Sato, Masami Ito, Mutsumi Kobayashi, Akitoshi Nakashima, Shinichiro Wada, Takashi Onda, Nobuya Unno, Yutaka Osuga
ABSTRACT
In endometriosis, M2 macrophages (MΦ) are dominant and promote the development of endometriosis lesions. However, the factor(s) which induces M2 MΦ are unknown. In the present study, we focused on interleukin (IL)-33, known as an alarmin and investigated its expression and its role in endometriosis, especially from the point of the relevance with MΦ. The expression of IL-33 in endometriosis lesions was examined by immunohistochemistry. The cystic fluid of ovarian cysts/tumors was obtained and used to measure IL-33 concentration. Endometriotic stromal cells (ESC) and MΦ derived from patients were used for in vitro experiments. IL-33 was detected in the epithelium and stromal cells of endometriotic lesions. The mean IL-33 concentration in the cystic fluid of endometriomas was significantly higher than that in non-endometriomas (2.2 ng/ml vs. 0.02 ng/ml, P < 0.01). IL-1β induced IL-33 mRNA expression in ESC via p38 MAPK activation. With IL-33 stimulation, peritoneal MΦ polarized to M2 MΦ and produced IL-1β mRNA with a 2.2-fold increase, which was negated with soluble ST2, a decoy receptor of IL-33. IL-33, derived from endometriotic lesions, stimulated MΦ to produce IL-1β, which results in increasing IL-33 production in ESC. This cycle may continue to exacerbate the endometriotic lesions.
Sigma-Aldrich
Anti-IL33 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.