  • Search Results
  • Sugar-Induced Obesity and Insulin Resistance Are Uncoupled from Shortened Survival in Drosophila.
Sugar-Induced Obesity and Insulin Resistance Are Uncoupled from Shortened Survival in Drosophila.
Cell metabolism (2020-03-21)
Esther van Dam, Lucie A G van Leeuwen, Eliano Dos Santos, Joel James, Lena Best, Claudia Lennicke, Alec J Vincent, Georgios Marinos, Andrea Foley, Marcela Buricova, Joao B Mokochinski, Holger B Kramer, Wolfgang Lieb, Matthias Laudes, Andre Franke, Christoph Kaleta, Helena M Cochemé
ABSTRACT
High-sugar diets cause thirst, obesity, and metabolic dysregulation, leading to diseases including type 2 diabetes and shortened lifespan. However, the impact of obesity and water imbalance on health and survival is complex and difficult to disentangle. Here, we show that high sugar induces dehydration in adult Drosophila, and water supplementation fully rescues their lifespan. Conversely, the metabolic defects are water-independent, showing uncoupling between sugar-induced obesity and insulin resistance with reduced survival in vivo. High-sugar diets promote accumulation of uric acid, an end-product of purine catabolism, and the formation of renal stones, a process aggravated by dehydration and physiological acidification. Importantly, regulating uric acid production impacts on lifespan in a water-dependent manner. Furthermore, metabolomics analysis in a human cohort reveals that dietary sugar intake strongly predicts circulating purine levels. Our model explains the pathophysiology of high-sugar diets independently of obesity and insulin resistance and highlights purine metabolism as a pro-longevity target.
Sigma-Aldrich
8-Hydroxypyrene-1,3,6-trisulfonic acid trisodium salt, ≥96%
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
D-(−)-Fructose, ≥99%
Sigma-Aldrich
Allantoin, ≥98.0% (N)
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Bromocresol Purple, Technical grade
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Adenine, ≥99%
Sigma-Aldrich
Bromophenol Blue sodium salt
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Methyl viologen dichloride hydrate, 98%
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Sigma-Aldrich
Mineral oil, heavy
Sigma-Aldrich
CelLytic M, Cell Lysis Reagent, Suitable for Mammalian cell lysis and protein solubilization.
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Propionic acid, ≥99.5%
Sigma-Aldrich
Anti-AGE (Advanced Glycated End-products) Antibody, serum, Chemicon®