  • Search Results
  • Versatile whole-organ/body staining and imaging based on electrolyte-gel properties of biological tissues.
Versatile whole-organ/body staining and imaging based on electrolyte-gel properties of biological tissues.
Nature communications (2020-04-29)
Etsuo A Susaki, Chika Shimizu, Akihiro Kuno, Kazuki Tainaka, Xiang Li, Kengo Nishi, Ken Morishima, Hiroaki Ono, Koji L Ode, Yuki Saeki, Kazunari Miyamichi, Kaoru Isa, Chihiro Yokoyama, Hiroki Kitaura, Masako Ikemura, Tetsuo Ushiku, Yoshihiro Shimizu, Takashi Saito, Takaomi C Saido, Masashi Fukayama, Hirotaka Onoe, Kazushige Touhara, Tadashi Isa, Akiyoshi Kakita, Mitsuhiro Shibayama, Hiroki R Ueda
ABSTRACT
Whole-organ/body three-dimensional (3D) staining and imaging have been enduring challenges in histology. By dissecting the complex physicochemical environment of the staining system, we developed a highly optimized 3D staining imaging pipeline based on CUBIC. Based on our precise characterization of biological tissues as an electrolyte gel, we experimentally evaluated broad 3D staining conditions by using an artificial tissue-mimicking material. The combination of optimized conditions allows a bottom-up design of a superior 3D staining protocol that can uniformly label whole adult mouse brains, an adult marmoset brain hemisphere, an ~1 cm3 tissue block of a postmortem adult human cerebellum, and an entire infant marmoset body with dozens of antibodies and cell-impermeant nuclear stains. The whole-organ 3D images collected by light-sheet microscopy are used for computational analyses and whole-organ comparison analysis between species. This pipeline, named CUBIC-HistoVIsion, thus offers advanced opportunities for organ- and organism-scale histological analysis of multicellular systems.
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ~99%
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Mineral oil, light oil, BioReagent, suitable for mouse embryo cell culture
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from salmon testes
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
N,N,N′,N′-Tetrakis(2-Hydroxypropyl)ethylenediamine, 98%
Sigma-Aldrich
CAPSO, ≥99% anhydrous basis (titration)
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
(+)-MK-801 hydrogen maleate, powder
Sigma-Aldrich
Anti-NeuN Antibody (rabbit), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, Alexa Fluor®488 conjugated, clone A60, Chemicon®, from mouse
Sigma-Aldrich
trans-1-Acetyl-4-hydroxy-L-proline, ≥98%