US
EN
Applications
Products
Services
Support
Home
Search Results
Matrix effect?corrected liquid chromatography/tandem mass-spectrometric method for determining acylcarnitines in human urine
Matrix effect?corrected liquid chromatography/tandem mass-spectrometric method for determining acylcarnitines in human urine
Clinica Chimica Acta; International Journal of Clinical Chemistry (2017)
Abe K, et al.