US
EN
Applications
Products
Services
Support
Home
Search Results
Polymerization of 3, 6-dibromo-9-(2, 3-epoxypropyl) carbazole with triphenylcarbenium salts
Polymerization of 3, 6-dibromo-9-(2, 3-epoxypropyl) carbazole with triphenylcarbenium salts
European Polymer Journal (1992)
Gravzulevivcius JV, et al.