  • Search Results
  • SARS-CoV-2 Reverse Genetics Reveals a Variable Infection Gradient in the Respiratory Tract.
SARS-CoV-2 Reverse Genetics Reveals a Variable Infection Gradient in the Respiratory Tract.
Cell (2020-06-12)
Yixuan J Hou, Kenichi Okuda, Caitlin E Edwards, David R Martinez, Takanori Asakura, Kenneth H Dinnon, Takafumi Kato, Rhianna E Lee, Boyd L Yount, Teresa M Mascenik, Gang Chen, Kenneth N Olivier, Andrew Ghio, Longping V Tse, Sarah R Leist, Lisa E Gralinski, Alexandra Schäfer, Hong Dang, Rodney Gilmore, Satoko Nakano, Ling Sun, M Leslie Fulcher, Alessandra Livraghi-Butrico, Nathan I Nicely, Mark Cameron, Cheryl Cameron, David J Kelvin, Aravinda de Silva, David M Margolis, Alena Markmann, Luther Bartelt, Ross Zumwalt, Fernando J Martinez, Steven P Salvatore, Alain Borczuk, Purushothama R Tata, Vishwaraj Sontake, Adam Kimple, Ilona Jaspers, Wanda K O'Neal, Scott H Randell, Richard C Boucher, Ralph S Baric
ABSTRACT
The mode of acquisition and causes for the variable clinical spectrum of coronavirus disease 2019 (COVID-19) remain unknown. We utilized a reverse genetics system to generate a GFP reporter virus to explore severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) pathogenesis and a luciferase reporter virus to demonstrate sera collected from SARS and COVID-19 patients exhibited limited cross-CoV neutralization. High-sensitivity RNA in situ mapping revealed the highest angiotensin-converting enzyme 2 (ACE2) expression in the nose with decreasing expression throughout the lower respiratory tract, paralleled by a striking gradient of SARS-CoV-2 infection in proximal (high) versus distal (low) pulmonary epithelial cultures. COVID-19 autopsied lung studies identified focal disease and, congruent with culture data, SARS-CoV-2-infected ciliated and type 2 pneumocyte cells in airway and alveolar regions, respectively. These findings highlight the nasal susceptibility to SARS-CoV-2 with likely subsequent aspiration-mediated virus seeding to the lung in SARS-CoV-2 pathogenesis. These reagents provide a foundation for investigations into virus-host interactions in protective immunity, host susceptibility, and virus pathogenesis.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Millipore
Millicell Cell Culture Insert, 12 mm, hydrophilic PTFE, 0.4 µm, Hydrophilic PTFE cell culture insert with pore size of 0.4 µm used in a 24-well plate for Cell Attachment, Cell Culture, Cell Differentiation, Fluorescence & Low Protein Binding.
Sigma-Aldrich
Anti-Tubulin Antibody, clone YL1/2, clone YL1/2, Chemicon®, from rat
Sigma-Aldrich
Anti-Clara Cell Secretory Protein Antibody, serum, Upstate®
Sigma-Aldrich
Galacto-N-biose
Sigma-Aldrich
Anti-Uteroglobin, serum, from goat