  • Search Results
  • NPM1 upregulates the transcription of PD-L1 and suppresses T cell activity in triple-negative breast cancer.
NPM1 upregulates the transcription of PD-L1 and suppresses T cell activity in triple-negative breast cancer.
Nature communications (2020-04-05)
Ge Qin, Xin Wang, Shubiao Ye, Yizhuo Li, Miao Chen, Shusen Wang, Tao Qin, Changlin Zhang, Yixin Li, Qian Long, Huabin Hu, Dingbo Shi, Jiaping Li, Kai Zhang, Qinglian Zhai, Yanlai Tang, Tiebang Kang, Ping Lan, Fangyun Xie, Jianjun Lu, Wuguo Deng
ABSTRACT
Programmed cell death protein-1 (PD-1)/programmed cell death ligand-1 (PD-L1) interaction plays a crucial role in tumor-associated immune escape. Here, we verify that triple-negative breast cancer (TNBC) has higher PD-L1 expression than other subtypes. We then discover that nucleophosmin (NPM1) binds to PD-L1 promoter specifically in TNBC cells and activates PD-L1 transcription, thus inhibiting T cell activity in vitro and in vivo. Furthermore, we demonstrate that PARP1 suppresses PD-L1 transcription through its interaction with the nucleic acid binding domain of NPM1, which is required for the binding of NPM1 at PD-L1 promoter. Consistently, the PARP1 inhibitor olaparib elevates PD-L1 expression in TNBC and exerts a better effect with anti-PD-L1 therapy. Together, our research has revealed NPM1 as a transcription regulator of PD-L1 in TNBC, which could lead to potential therapeutic strategies to enhance the efficacy of cancer immunotherapy.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Millipore
Streptavidin−Agarose from Streptomyces avidinii, buffered aqueous suspension
Millipore
ANTI-FLAG® M1 Agarose Affinity Gel