  • Search Results
  • Histone deacetylase HDA-1 modulates mitochondrial stress response and longevity.
Histone deacetylase HDA-1 modulates mitochondrial stress response and longevity.
Nature communications (2020-09-17)
Li-Wa Shao, Qi Peng, Mingyue Dong, Kaiyu Gao, Yumei Li, Yi Li, Chuan-Yun Li, Ying Liu
ABSTRACT
The ability to detect, respond and adapt to mitochondrial stress ensures the development and survival of organisms. Caenorhabditis elegans responds to mitochondrial stress by activating the mitochondrial unfolded protein response (UPRmt) to buffer the mitochondrial folding environment, rewire the metabolic state, and promote innate immunity and lifespan extension. Here we show that HDA-1, the C. elegans ortholog of mammalian histone deacetylase (HDAC) is required for mitochondrial stress-mediated activation of UPRmt. HDA-1 interacts and coordinates with the genome organizer DVE-1 to induce the transcription of a broad spectrum of UPRmt, innate immune response and metabolic reprogramming genes. In rhesus monkey and human tissues, HDAC1/2 transcript levels correlate with the expression of UPRmt genes. Knocking down or pharmacological inhibition of HDAC1/2 disrupts the activation of the UPRmt and the mitochondrial network in mammalian cells. Our results underscore an evolutionarily conserved mechanism of HDAC1/2 in modulating mitochondrial homeostasis and regulating longevity.
Product Description
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Sodium butyrate, 98%