  • Search Results
  • Crypt residing bacteria and proximal colonic carcinogenesis in a mouse model of Lynch syndrome.
Crypt residing bacteria and proximal colonic carcinogenesis in a mouse model of Lynch syndrome.
International journal of cancer (2020-05-01)
Michaela Lang, Maximilian Baumgartner, Aleksandra Rożalska, Adrian Frick, Alessandra Riva, Michael Jarek, David Berry, Christoph Gasche
ABSTRACT
Colorectal cancer is a multifactorial disease involving inherited DNA mutations, environmental factors, gut inflammation and intestinal microbiota. Certain germline mutations within the DNA mismatch repair system are associated with Lynch syndrome tumors including right-sided colorectal cancer with mucinous phenotype and presence of an inflammatory infiltrate. Such tumors are more often associated with bacterial biofilms, which may contribute to disease onset and progression. Inflammatory bowel diseases are also associated with colorectal cancer and intestinal dysbiosis. Herein we addressed the question, whether inflammation can aggravate colorectal cancer development under mismatch repair deficiency. MSH2loxP/loxP Vill-cre mice were crossed into the IL-10-/- background to study the importance of inflammation and mucosal bacteria as a driver of tumorigenesis in a Lynch syndrome mouse model. An increase in large bowel tumorigenesis was found in double knockout mice both under conventional housing and under specific pathogen-free conditions. This increase was mostly due to the development of proximal tumors, a hotspot for tumorigenesis in Lynch syndrome, and was associated with a higher degree of inflammation. Additionally, bacterial invasion into the mucus of tumor crypts was observed in the proximal tumors. Inflammation shifted fecal and mucosal microbiota composition and was associated with enrichment in Escherichia-Shigella as well as Akkermansia, Bacteroides and Parabacteroides genera in fecal samples. Tumor-bearing double knockout mice showed a similar enrichment for Escherichia-Shigella and Parabacteroides. Lactobacilli, Lachnospiraceae and Muribaculaceae family members were depleted upon inflammation. In summary, chronic inflammation aggravates colonic tumorigenesis under mismatch repair deficiency and is associated with a shift in microbiota composition.
Product Description
Millipore
Gram Staining Kit, for microscopy, used for differentiation of bacteria on the basis of their gram nature.
Sigma-Aldrich
Tofacitinib citrate, ≥98% (HPLC)