  • Search Results
  • X-ray induced photodynamic therapy (PDT) with a mitochondria-targeted liposome delivery system.
X-ray induced photodynamic therapy (PDT) with a mitochondria-targeted liposome delivery system.
Journal of nanobiotechnology (2020-06-12)
Xuefan Gu, Chao Shen, Hua Li, Ewa M Goldys, Wei Deng
ABSTRACT
In this study, we constructed multifunctional liposomes with preferentially mitochondria-targeted feature and gold nanoparticles-assisted synergistic photodynamic therapy. We systemically investigated the in vitro X-ray triggered PDT effect of these liposomes on HCT 116 cells including the levels of singlet oxygen, mitochondrial membrane potential, cell apoptosis/necrosis and the expression of apoptosis-related proteins. The results corroborated that synchronous action of PDT and X-ray radiation enhance the generation of cytotoxic reactive oxygen species produced from the engineered liposomes, causing mitochondrial dysfunction and increasing the levels of apoptosis.
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
McCoy′s 5A Medium, Modified, with sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Verteporfin, ≥94% (HPLC)
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
(4-Carboxybutyl)triphenylphosphonium bromide, 98%