  • Search Results
  • Pseudorabies virus infection inhibits stress granules formation via dephosphorylating eIF2α.
Pseudorabies virus infection inhibits stress granules formation via dephosphorylating eIF2α.
Veterinary microbiology (2020-08-10)
Shengkui Xu, Dongjie Chen, Dengjin Chen, Qianlin Hu, Lei Zhou, Xinna Ge, Jun Han, Xin Guo, Hanchun Yang
ABSTRACT
Pseudorabies virus (PRV) is one of the most notorious pathogens in the global pig industry. During infection, viruses may evolve various strategies, such as modulating stress granules (SGs) formation, to create an optimal surroundings for viral replication. However, the interplay between PRV infection and SGs formation remains largely unknown. Here we showed that PRV infection markedly blocked SGs formation induced by sodium arsenate (AS) and DL-Dithiothreitol (DTT). Accordantly, the phosphorylation of eIF2α was markedly inhibited in PRV-infected cells, although two eIF2α kinases double-stranded RNA-activated protein kinase (PKR) and PKR-like ER kinase (PERK) were activated during PRV infection. Furthermore, we also found that the dephosphorylation of eIF2α occurred at the early stage of virus infection but without the elevated production of GADD34 and PP1. Moreover, inhibition of PP1 activity by salubrinal could counteract PRV-mediated eIF2α dephosphorylation partially and inhibit virus replication. Our results revealed that, on the one hand, PRV infection activated eIF2α kinases PKR (latter inhibited) and PERK, and on the other hand, PRV encoded-functions dephosphorylated eIF2α and inhibited SGs formation to facilitate virus replication.
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium (meta)arsenite, ≥90%
Sigma-Aldrich
PP1, ≥98% (HPLC)