  • Search Results
  • LINC00473 functions as an oncogene and predicts poor prognosis in pancreatic cancer via the cAMP/β-catenin axis.
LINC00473 functions as an oncogene and predicts poor prognosis in pancreatic cancer via the cAMP/β-catenin axis.
European review for medical and pharmacological sciences (2020-06-05)
X-F Hao, Y-X Zhang, X-X Wang, W Sun, J Zhao, G-R Chen
ABSTRACT
To investigate the expression and function of LINC00463 in pancreatic cancer (PC), and to demonstrate the relationship between LINC00473 expression and clinical pathological characteristics and prognosis of PC. Expressions of LINC00473 in PC tissues and cell lines were detected using quantitative Real Time-Polymerase Chain Reaction (qRT-PCR). LINC00473 siRNA was synthesized to knock down the LINC00473 expression in PANC-1 cells. Proliferation, invasion, and migration abilities of experimental cells were analyzed using cell counting kit-8 (CCK-8) assay and transwell assay, respectively. cAMP activity was detected and protein expression of β-catenin was measured to explain the underlying mechanism of LINC00473 in PC. The prognosis and clinical pathological features of PC patients were illustrated. LINC00473 was highly expressed in PC tissues and cells. Higher level of LINC00473 was relative with larger tumor size, worse tumor node metastasis (TNM) stage, worse tumor differentiation, higher rates of perineural invasion, and lymphatic invasion. Knockdown of LINC00473 significantly inhibited cell growth, invasion, and migration of PANC-1 cells. LINC00473 activated cAMP and then promoted the phosphorylation of β-catenin to promote the progression of PC. Furthermore, high expression of LINC00473 and β-catenin remarkedly indicated poor prognosis of PC patients. LINC00473 was upregulated in PC tissues and cells, indicating a poor prognosis and clinical pathological features of PC. It promoted PC progression via activating the cAMP/β-catenin axis, which provided a novel target for the prediction for PC diagnosis, biological therapy, and prognosis.
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1