  • Search Results
  • Knockout of reactive astrocyte activating factors slows disease progression in an ALS mouse model.
Knockout of reactive astrocyte activating factors slows disease progression in an ALS mouse model.
Nature communications (2020-07-29)
Kevin A Guttenplan, Maya K Weigel, Drew I Adler, Julien Couthouis, Shane A Liddelow, Aaron D Gitler, Ben A Barres
ABSTRACT
Reactive astrocytes have been implicated in the pathogenesis of neurodegenerative diseases, including a non-cell autonomous effect on motor neuron survival in ALS. We previously defined a mechanism by which microglia release three factors, IL-1α, TNFα, and C1q, to induce neurotoxic astrocytes. Here we report that knocking out these three factors markedly extends survival in the SOD1G93A ALS mouse model, providing evidence for gliosis as a potential ALS therapeutic target.
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture