  • Search Results
  • Acute Valproate Exposure Induces Sex-Specific Changes in Steroid Hormone Metabolism in the Cerebral Cortex of Juvenile Mice.
Acute Valproate Exposure Induces Sex-Specific Changes in Steroid Hormone Metabolism in the Cerebral Cortex of Juvenile Mice.
Neurochemical research (2020-07-01)
Sung-Hee Cho, Jung Hoon Chai, Sung-Youn Chang, Soon Ae Kim
ABSTRACT
Valproic acid (VPA), an antiepileptic and mood stabilizer, modulates neurotransmission and gene expression by inhibiting histone deacetylase activity. It is reported that VPA may affects the steroid hormone level. In this study, VPA-induced acute metabolic alterations were investigated using liquid chromatography-tandem mass spectrometry in prepubertal mice brain. In VPA-treated (400 mg/kg in saline solution, intraperitoneal) mice, cortisol levels were increased (female: P < 0.004, male: P < 0.003) and 17β-estradiol levels were decreased (Both P < 0.03). Furthermore, in the VPA-treated male mice, dihydrotestosterone levels were increased (P < 0.02) and testosterone were decreased (P < 0.002). The 4-hydroxylase activity was upregulated in the female VPA-treated mice (P < 0.01) and the 5α-reductase activity was increased in the male VPA-treated mice (P < 0.003). These results indicate sex specific differences in VPA-induced steroid metabolism in the brain cortex.
Sigma-Aldrich
5β-Pregnan-3α-ol-20-one
Sigma-Aldrich
5α-Pregnan-3α-ol-20-one, solid
Sigma-Aldrich
21-Deoxycortisol
Sigma-Aldrich
16-Epiestriol, ≥93% (HPLC)
Sigma-Aldrich
trans-Androsterone
Sigma-Aldrich
Aldosterone, ≥95% (HPLC)