  • Search Results
  • Modulatory effect of methanandamide on gastric vagal afferent satiety signals depends on nutritional status.
Modulatory effect of methanandamide on gastric vagal afferent satiety signals depends on nutritional status.
The Journal of physiology (2020-04-03)
Stewart Christie, Rebecca O'Rielly, Hui Li, Maria Nunez-Salces, Gary A Wittert, Amanda J Page
ABSTRACT
Gastric vagal afferent responses to tension are dampened in high fat diet-induced obesity. Endocannabinoids are known to dose-dependently inhibit and excite gastric vagal afferents but their effect on gastric vagal afferents in diet-induced obesity are unknown. In individual gastric vagal afferent neurons of diet-induced obese mice the co-expression of components of the endocannabinoid system, including CB1, GHSR, TRPV1 and FAAH, was increased compared with lean mice. In high fat diet-induced obese mice, methanandamide only inhibited gastric vagal afferent responses to tension, possibly due to the observed change in the balance of receptors, hormones and breakdown enzymes in this system. Collectively, these data suggest that endocannabinoid signalling, by gastric vagal afferents, is altered in diet-induced obesity which may impact satiety and gastrointestinal function. Gastric vagal afferents (GVAs) play a role in appetite regulation. The endocannabinoid anandamide (AEA) dose-dependently inhibits and excites tension-sensitive GVAs. However, it is also known that high fat diet (HFD) feeding alters GVA responses to stretch. The aim of this study was to determine the role of AEA in GVA signalling in lean and HFD-induced obese mice. Male C57BL/6 mice were fed (12 weeks) a standard laboratory diet (SLD) or HFD. Protein and mRNA expression of components of the cannabinoid system was determined in individual GVA cell bodies and the gastric mucosa. An in vitro GVA preparation was used to assess the effect of methanandamide (mAEA) on tension-sensitive GVAs and the second messenger pathways involved. In individual GVA cell bodies, cannabinoid 1 (CB1) and ghrelin (GHSR) receptor mRNA was higher in HFD mice than SLD mice. Conversely, gastric mucosal AEA and ghrelin protein levels were lower in HFD mice than SLD mice. In SLD mice, mAEA exerted dose-dependent inhibitory and excitatory effects on tension-sensitive GVAs. Only an inhibitory effect of mAEA was observed in HFD mice. The excitatory effect of mAEA was dependent on CB1, transient receptor potential vanilloid 1 (TRPV1) and the protein kinase C. Conversely, the inhibitory effect was dependent on CB1, growth hormone secretagogue receptor, TRPV1 and the protein kinase A. Endocannabinoids, acting through CB1 and TRPV1, have a pivotal role in modulating GVA satiety signals depending on the second messenger pathway utilised. In HFD mice only an inhibitory effect was observed. These changes may contribute to the development and/or maintenance of obesity.
Sigma-Aldrich
Arachidonylethanolamide, ≥97.0% (TLC), oil
Sigma-Aldrich
Rimonabant hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
AMG 9810, ≥98% (HPLC), powder
Sigma-Aldrich
Protein Kinase A Inhibitor Fragment 6-22 amide, ≥97% (HPLC)
Sigma-Aldrich
Bisindolylmaleimide II, ≥97% (Mixture of 2 isomers)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.