  • Search Results
  • The Bioinformatic and In Vitro Studies of Clostridioides Difficile Aminopeptidase M24 Revealed the Immunoreactive KKGIK Peptide.
The Bioinformatic and In Vitro Studies of Clostridioides Difficile Aminopeptidase M24 Revealed the Immunoreactive KKGIK Peptide.
Cells (2020-05-13)
Katarzyna Pacyga, Agnieszka Razim, Gayane Martirosian, Małgorzata Aptekorz, Andrzej Szuba, Andrzej Gamian, Andrzej Myc, Sabina Górska
ABSTRACT
Clostridioides difficile (CD) is a Gram-positive pathogen responsible for CD-associated disease (CDAD), which is characterized by symptoms ranging from mild diarrhea to pseudomembranous colitis. This work is an attempt to respond to the need of novel methods for CD infection (CDI) prevention, since the number of CDI cases is still rising. A bioinformatics approach was applied to design twenty-one peptides consisting of in silico predicted linear B-cell and T-cell epitopes of aminopeptidase M24 from CD. These peptides were mapped for epitopes exploiting PEPSCAN procedure and using sera obtained from CD infected patients, umbilical cord blood, and healthy volunteers. Two new CD epitopes, 131KKGIK135 and 184KGTSTHVIT192, were identified and characterized. Immunoreactivity of the synthetic biotinylated 131KKGIK135 epitope was significantly higher compared to 184KGTSTHVIT192 epitope in Enzyme-Linked Immunosorbent Assay (ELISA) with umbilical cord blood and CDI patients' sera. Hereafter, the conjugate of bovine serum albumin and epitope 131KKGIK135 was evaluated in vitro on lung epithelial cell line. In vitro, a significant induction of IL-6 by conjugate was observed, thereby we postulate that this new 131KKGIK135 epitope possesses immunostimulating properties suggesting possibility of its use in a vaccine against Clostridioides difficile.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Anti-Human IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.