  • Search Results
  • Magnitude and direction of vesicle dynamics in growing pollen tubes using spatiotemporal image correlation spectroscopy and fluorescence recovery after photobleaching.
Magnitude and direction of vesicle dynamics in growing pollen tubes using spatiotemporal image correlation spectroscopy and fluorescence recovery after photobleaching.
Plant physiology (2008-05-30)
Jérôme Bove, Benoit Vaillancourt, Jens Kroeger, Peter K Hepler, Paul W Wiseman, Anja Geitmann
ABSTRACT
The delivery of cell wall material and membrane to growing plant cell surfaces requires the spatial and temporal coordination of secretory vesicle trafficking. Given the small size of vesicles, their dynamics is difficult to quantify. To quantitatively analyze vesicle dynamics in growing pollen tubes labeled with the styryl dye FM1-43, we applied spatiotemporal correlation spectroscopy on time-lapse series obtained with high-speed confocal laser scanning microscopy recordings. The resulting vector maps revealed that vesicles migrate toward the apex in the cell cortex and that they accumulate in an annulus-shaped region adjacent to the extreme tip and then turn back to flow rearward in the center of the tube. Fluorescence recovery after photobleaching confirmed vesicle accumulation in the shoulder of the apex, and it revealed that the extreme apex never recovers full fluorescence intensity. This is consistent with endocytotic activity occurring in this region. Fluorescence recovery after photobleaching analysis also allowed us to measure the turnover rate of the apical vesicle population, which was significantly more rapid than the theoretical rate computed based on requirements for new cell wall material. This may indicate that a significant portion of the vesicles delivered to the apex does not succeed in contacting the plasma membrane for delivery of their contents. Therefore, we propose that more than one passage into the apex may be needed for many vesicles before they fuse to the plasma membrane and deliver their contents.
Sigma-Aldrich
Agarose, low gelling temperature, BioReagent, for molecular biology
Sigma-Aldrich
Agarose, low gelling temperature, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Agarose, low gelling temperature, Type VII-A