  • Search Results
  • GRIM-19 is essential for maintenance of mitochondrial membrane potential.
GRIM-19 is essential for maintenance of mitochondrial membrane potential.
Molecular biology of the cell (2008-02-22)
Hao Lu, Xinmin Cao
ABSTRACT
GRIM-19 was found to copurify with complex I of mitochondrial respiratory chain and subsequently was demonstrated to be involved in complex I assembly and activity. To further understand its function in complex I, we dissected its functional domains by generating a number of deletion, truncation, and point mutants. The mitochondrial localization sequences were located at the N-terminus. Strikingly, deletion of residues 70-80, 90-100, or the whole C-terminal region (70-144) led to a loss of mitochondrial transmembrane potential (DeltaPsim). However, similar deletions of another two complex I subunits, NDUFA9 and NDUFS3, did not show such effect. We also found that deletion of the last 10 residues affected GRIM-19's ability to be assembled to complex I. We constructed a dominant-negative mutant containing the N-terminal 60 and the last C-terminal 10 residues, which could be assembled into complex I, but failed to maintain normal DeltaPsim. Cells overexpressing this mutant did not spontaneously undergo cell death, but were sensitized to apoptosis induced by cell death agents. Our results demonstrate that GRIM-19 is required for electron transfer activity of complex I, and disruption of DeltaPsim by GRIM-19 mutants enhances the cells' sensitivity to apoptotic stimuli.
Product Description
SAFC
RPMI-1640 Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Supelco
Rotenone, PESTANAL®, analytical standard