  • Search Results
  • Natural selection of FLT1 alleles and their association with malaria resistance in utero.
Natural selection of FLT1 alleles and their association with malaria resistance in utero.
Proceedings of the National Academy of Sciences of the United States of America (2008-09-10)
Atis Muehlenbachs, Michal Fried, Jeff Lachowitzer, Theonest K Mutabingwa, Patrick E Duffy
ABSTRACT
Placental malaria (PM) caused by Plasmodium falciparum contributes significantly to infant mortality in sub-Saharan Africa and is associated with pregnancy loss. We hypothesized that fetal genes that modify PM would be associated with fetal fitness. During PM, placental trophoblasts produce soluble fms-like tyrosine kinase 1 (sFlt1), also known as soluble VEGF receptor 1, an angiogenesis inhibitor associated with preeclampsia. Here we present a study examining the genotype of the fms-related tyrosine kinase 1 (FLT1) 3' UTR in Tanzanian mother-infant pairs. First-time mothers suffer the most PM, and newborn FLT1 genotype distribution differed by birth order, with newborns of first-time mothers outside of Hardy-Weinberg equilibrium (HWE) during peak PM season. Among first-time but not other mothers, maternal FLT1 genotype was associated with a history of prior pregnancy loss. During PM, newborn FLT1 genotype was associated with low birth weight and placental inflammatory gene expression. FLT1 genotype was also associated with Flt1 levels among study subjects and in vitro. Thus, FLT1 variants confer fetal fitness in utero and are associated with the maternal immune response during PM. This indicates that FLT1 is under natural selection in a malaria endemic area and that human exposure to malaria can influence the evolutionary genetics of the maternal-fetal relationship.
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
SAFC
RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture