  • Search Results
  • The enzyme mechanism of nitrite reductase studied at single-molecule level.
The enzyme mechanism of nitrite reductase studied at single-molecule level.
Proceedings of the National Academy of Sciences of the United States of America (2008-02-28)
Sofya Kuznetsova, Gerhild Zauner, Thijs J Aartsma, Hans Engelkamp, Nikos Hatzakis, Alan E Rowan, Roeland J M Nolte, Peter C M Christianen, Gerard W Canters
ABSTRACT
A generic method is described for the fluorescence "readout" of the activity of single redox enzyme molecules based on Förster resonance energy transfer from a fluorescent label to the enzyme cofactor. The method is applied to the study of copper-containing nitrite reductase from Alcaligenes faecalis S-6 immobilized on a glass surface. The parameters extracted from the single-molecule fluorescence time traces can be connected to and agree with the macroscopic ensemble averaged kinetic constants. The rates of the electron transfer from the type 1 to the type 2 center and back during turnover exhibit a distribution related to disorder in the catalytic site. The described approach opens the door to single-molecule mechanistic studies of a wide range of redox enzymes and the precise investigation of their internal workings.
Sigma-Aldrich
Atto 655, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Atto 655 NHS ester, BioReagent, suitable for fluorescence
Sigma-Aldrich
Atto 655-Biotin, BioReagent, suitable for fluorescence, ≥95.0% (HPCE)
Sigma-Aldrich
Atto 655 amine, suitable for fluorescence
Sigma-Aldrich
Atto 655-maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling rate)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.