MilliporeSigma
Development of arterial blood supply in experimental liver metastases.
The American journal of pathology (2009-07-04)
Katalin Dezso, Edina Bugyik, Veronika Papp, Viktória László, Balázs Döme, József Tóvári, József Tímár, Péter Nagy, Sándor Paku
ABSTRACT
In this study, we present a mechanism for the development of arterial blood supply in experimental liver metastases. To analyze the arterialization process of experimental liver metastases, we elucidated a few key questions regarding the blood supply of hepatic lobules in mice. The microvasculature of the mouse liver is characterized by numerous arterioportal anastomoses and arterial terminations at the base of the lobules. These terminations supply one hepatic microcirculatory subunit per lobule, which we call an arterial hepatic microcirculatory subunit (aHMS). The process of arterialization can be divided into the following steps: 1) distortion of the aHMS by metastasis; 2) initial fusion of the sinusoids of the aHMS at the tumor parenchyma interface; 3) fusion of the sinusoids located at the base of the aHMSs, which leads to the disruption of the vascular sphincter (burst pipe); 4) incorporation of the dilated artery and the fused sinusoids into the tumor; and 5) further development of the tumor vasculature (arterial tree) by proliferation, remodeling, and continuous incorporation of fused sinusoids at the tumor-parenchyma interface. This process leads to the inevitable arterialization of liver metastases above the 2000- to 2500-mum size, regardless of the origin and growth pattern of the tumor.
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, from USDA approved countries, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, γ-irradiated by SER-TAIN process, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, γ-irradiated, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, Heat Inactivated, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Canada origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USDA approved, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Australia origin, suitable for, USDA approved, sterile-filtered, suitable for cell culture, suitable for hybridoma
SAFC
Fetal Bovine Serum, Australia origin, γ-irradiated, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Charcoal Stripped, sterile-filtered, suitable for cell culture